Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPHK2 Monoclonal Antibody (M532)
GREENER_CHOICE

Catalog No. PIMA547737
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA547737 100 μL
1 options

Catalog No. PIMA547737

Supplier: Invitrogen™ MA547737

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody detects 70 kDa* proteins corresponding to the molecular mass of SK2 on SDS-PAGE immunoblots of human recombinant SK2 and endogenous SK2 in human HeLa cells.

SPHK2, along with SPHK1, catalyzes the phosphorylation of sphingosine producing the signaling molecule sphingosine-1-phosphate (S1P). S1P signals through G-protein coupled receptors and controls diverse cellular processes. Inhibitors of SPHKs are viewed as potential therapeutics for the treatment of cancer. SPHK2 is an important signaling molecule with intra- and extracellular functions and has been implicated in regulating cell differentiation, calcium mobilization from intracellular stores, and apoptosis. The cell surface receptors for S1P are the EDG family of G protein-coupled receptors (S1P Receptors). These receptors couple to multiple G proteins (e.g. S1P1 couples to Gi whereas S1P2 and S1P3 couple to Gq, G13 in addition to Gi) and regulate a extremely wide raange of cellular events including cell motility, survival, apoptosis, migration and cell-cell interaction. Important roles for S1P have also been reported in regulation of cardiogenesis, vascular maturation, oocyte survival, immune cell trafficking, cells of the neuronal system and bone cells. S1P levels are regulated by the activity of Sphk (Sphk1 and Sphk2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPHK2
Applications Western Blot
Classification Monoclonal
Clone M532
Concentration ∼1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene Sphk2
Gene Accession No. Q9JIA7, Q9NRA0
Gene Alias C76851; SK 2; SK2; SK-2; Sphingosine kinase 2; sphingosine kinase type 2; Sphk2; SPK 2; SPK-2; Spk2, Sk2
Gene Symbols Sphk2
Host Species Mouse
Immunogen Clone M532 was generated from a SK2 (N-terminal region) synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminal region of human SK2. This peptide sequence is highly conserved in rat and mouse SK2 proteins, and has no homology to SK1.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308589, 56632, 56848
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.