Learn More
Cayman Chemical SphngosIn1PhosphAT Lyase 1mg
SDP
Supplier:  Cayman Chemical 132381 MG
Encompass_Preferred
A matrix metalloproteinase inhibitor inhibits MMP-2 and MMP-9 (IC50s 4 and 20 UM respectively) prevents 90 of tumor cell invasion across matrigel-coated membranes in vitro at 100 UM
Catalog No. 50-295-1323
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.