Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Sphygmomanometer
SDPSCI_ED

Catalog No. S15678 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Size:
Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S15678 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is intended for educational purposes only.
Catalog No. S15678 Supplier United Scientific™ Supplier No. SPHY01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is intended for educational purposes only.

This student blood pressure set has a self-adjusting cuff with gauge holder

  • This student blood pressure set has a self-adjusting cuff with gauge holder.
  • Includes a vinyl storage pouch.

Specifications

Display Analog
Size Standard
For Use With (Equipment) Blood Pressure Devices and Patient Monitors
Display Type Gauge
For Use With (Application) Finding Blood Pressure
Includes Vinyl Storage Pouch
Product Type Sphygmomanometer
Color Black and Green

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.