Learn More
Description
Key information: SPiDER-βGal Cellular Senescence Dye is a fluorogenic β-galactosidase substrate suitable for labeling live cells in culture and living tissues. It exhibits fluorescence upon reaction with the enzyme, remains inside cells by anchoring itself to intracellular proteins and provides single-cell resolution.
Used for: labeling and imaging lacZ reporters in living cells, tissues and model organisms. Also used to investigate senescence.
Application: Confocal microscopy, flow cytometry, fluorescence microscopy. Has high sensitivity for the detection of peritoneal ovarian cancer metastases in animal models and displays high constrast of tumor margins. Also used for investigating senescence of pancreatic cells and imaging adhesion G-protein-coupled receptors expressed in endothelial cells.
Properties and Photophysical Data: Excitation and emission maxima (λ) measured in 200 mM sodium phosphate buffer are 500-540 nm and 530-570 nm, respectively; activation ratio >650-fold.
Please refer to this protocol for further information on how to use this product.
Specifications
Specifications
| Product Line | Tocris |
| Quantity | 20 μg |
| Substrate Type | Beta-Gal Substrate |
| Storage Requirements | Store at +4°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
