Learn More
Justrite™ Polypropylene, Spigot Fitting, 1-1/8 in. Thread With 3/4 in. Sanitary Connect
Shop All Justrite Safety Group Products
Description
- With easy-grip lever to control continuous or manual flow rates
- Designed to withstand 100L pressure
- With closed cap
Specifications
Specifications
| Description | 1-1/8 in. Thread with 3/4 in. Sanitary Connect |
| For Use With (Equipment) | Carboys |
| Material | Polypropylene |
| No. per Pack | 1 Ea. |
| Product Type | Spigot Fitting |