Learn More
Pro Sci Inc SARS-CoV-2 (COVID-19) Spike RBD Antibody | 0.1mg
SDP
Supplier:  Pro Sci Inc 9087
Encompass_Preferred
Anti-SARS-CoV-2 (COVID-19) Spike RBD antibody (9087) was raised against a peptide corresponding to 19 amino acids near the carboxy terminus of SARS-CoV-2 (COVID-19) Spike glycoprotein RBD. The immunogen is located within the last 50 amino acids of SARS-CoV-2 (COVID-19) Spike protein RBD.
Catalog No. NC1836152
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.