Learn More
ACROBiosystems Bat coronavirus BM48-31/BGR/2008 Spike Trimer, His Tag (SPN-B52H9) is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag at the C-terminus. It contains AA Asn 14 - Pro 1199 (Accession # E0XI
Supplier:  ACROBiosystems SPNB52H950UG
Encompass_Preferred
It's been reported that SARS-CoV-2 can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
Catalog No. 50-225-3758