Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spike RBD Mouse, anti-SARS-CoV-2, Unconjugated, MP Biomedicals™

Catalog No. MP08L100002 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08L100002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP08L100002 Supplier MP Biomedicals, Inc Supplier No. 08L100002
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SARS-CoV-2 Spike RBD Mouse Monoclonal Antibody has been experimentally validated in ELISA, WB, IF assays.

  • The spike (S) glycoprotein of coronaviruses contains protrusions that will only bind to certain receptors on the host cell, which is essential for both host specificity and viral infectivity
  • The spike (S) glycoprotein of coronaviruses is known to be essential in the binding of the virus to the host cell at the advent of the infection process
  • Offers high specificity and potency against SARS-CoV-2 antigens in immunological and biochemical assays and consistency between lots
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spike RBD
Applications ELISA, Western Blot, Immunofluorescence
Classification Monoclonal
Concentration 3.49 mg/mL
Conjugate Unconjugated
Dilution ELISA: 1:37500 to 1:150000; Western Blot: 1:500 to 1:2000; Immunofluorescence: 1:50 to 1:200.
Formulation In PBS buffer with 0.02% sodium azide, 0.05% BSA, 50% glycerol, pH7.3 to 7.4.
Gene Alias S1-RBD protein, NCP-CoV RBD Protein, novel coronavirus RBD Protein, 2019-nCoV RBD Protein, S glycoprotein Subunit1 RBD Protein
Host Species Mouse
Immunogen Full-length SARS-CoV-2 Spike RBD protein
Purification Method Affinity Purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species SARS-CoV-2
Content And Storage -20°C
Form Liquid
Isotype IgG
Show More Show Less

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.