Learn More
ACROBiosystems This kit is developed for detecting SARS-CoV-2 Spike RBD in vaccine samples,witch is the low sensitivity version of Cat.No. RAS-A021.
Supplier:  ACROBiosystems RASA05096TESTS
Encompass_Preferred
The newly identified Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) has posed a serious threat to human health. A rapid and effective assay kit detecting the levels of SARS-CoV-2 Spike Protein RBD is urgently needed to accelerate the development of COVID-19 vaccines.
Catalog No. 50-223-1123
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.