Learn More
Pro Sci Inc SARS-CoV-2 (COVID-19) Spike S1 Antibody | 0.1mg
SDP
Supplier:  Pro Sci Inc 9083
Encompass_Preferred
Anti-SARS-CoV-2 (COVID-19) Spike S1 antibody (9083) was raised against a peptide corresponding to 16 amino acids near the amino terminus of SARS-CoV-2 (COVID-19) Spike S1 glycoprotein. The immunogen is located within the first 50 amino acids of SARS-CoV-2 (COVID-19) Spike S1 protein.
Catalog No. NC1836151
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.