Learn More
ACROBiosystems ACROBiosystems Biotinylated SARS-CoV-2 Spike Trimer (T19R, V70F, FR157-158Del, A222V, W258L, K417N, L452R, T478K, D614G, P681R, D950N), His,Avitag™
Supplier:  ACROBiosystems SPNC82EI200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (SPN-C82Ei) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 139.5 kDa.
Catalog No. NC2010474
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.