Learn More
New Pig Spill Kit in Small Wall - Mount Cabinet
SDP
Catalog No. 17987833 Shop All New Pig Corporation Products
Change view
Click to view available options
Type:
MRO Small Wall Mount Spill Kit
Catalog No. Type
17-987-833 MRO Small Wall Mount Spill Kit
1 options
Catalog No. 17-987-833 Supplier New Pig Supplier No. KIT215
Add to Cart
Add to Cart
Wall mountable kit allows you to locate your spill supplies right where you need them - so they're handy when you need them
  • The wall-mountable design allows you to place the kit exactly where spills are most likely to occur, ensuring a fast and efficient cleanup.
  • The cabinet is pre-stocked with a variety of absorbents and other spill response tools, so you're prepared to handle spills immediately.
  • The bright safety-yellow, steel cabinet is clearly labeled Spill Kit for quick identification in an emergency.
  • The sturdy, wall-mounted cabinet keeps your spill supplies neat, organized, and protected from damage or tampering.
  • A clear front panel lets you easily monitor the contents of the kit, so you know at a glance when supplies need to be restocked.
  • The kit is designed to absorb a wide range of liquids including oils, coolants, solvents, and water, making it a versatile solution for most spills.
Specifications
Type MRO Small Wall Mount Spill Kit
Weight Capacity Heavyweight
Weight (English) 39.6 lb.
Weight (Metric) 18 kg