Learn More
New Pig Spill-X™ Acid-Neutralizing Shaker Bottle
SDP
Catalog No. 17987832 Shop All New Pig Corporation Products
Catalog No. 17-987-832 Supplier New Pig Supplier No. GEN528
Add to Cart
Add to Cart
Neutralizes smaller spills for safer cleanup and handling. Great for labs
  • Quickly and safely neutralizes acid spills: The shaker bottle contains a proprietary agent that neutralizes and solidifies a variety of acids, making cleanup safer and easier.
  • Ideal for laboratory environments: Its compact and easy-to-use design makes it perfect for containing and treating small spills in labs and other areas where chemicals are used.
  • Reduces exposure and disposal costs: The granular base lets you use only what you need, minimizing waste and helping to reduce the overall cost of spill cleanup and disposal.
  • Turns hazardous spills into safe solids: The agent changes the pH of the acid, eliminating its corrosive properties and creating a solid, neutralized salt that is much easier to handle and dispose of.
  • Fast-acting and efficient: The free-flowing dry chemical allows for quick and precise application, speeding up spill cleanup and limiting worker exposure time.
  • Non-toxic and environmentally safe: The agent itself is a non-toxic dry chemical that is environmentally safe, helping to protect your workplace and the planet.