Learn More
New Pig SpillBlocker™ Tabletop Dike
SDP
Catalog No. 18389143 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: SpillBlocker™rade; Tabletop Dike
Catalog No.
18-389-143 Type: SpillBlocker™rade; Tabletop Dike
1 options
This item is not returnable. View return policy
Catalog No. 18-389-143 Supplier New Pig Supplier No. PLR229
Add to Cart
Add to Cart
This item is not returnable. View return policy
Contain small spills on workbenches or in labs with this liquid-proof, low-profile mini-barrier.
  • Contains benchtop spills without absorbing; ideal for small spills on smooth work surfaces
  • Polyurethane material resists water, oils and many chemicals High-visibility yellow color draws attention to spill for increased safety
  • Reusable dike cleans easily with soap and water
  • Comes in compact storage box for easy transport Cut to any length for a custom fit
  • Can also be used to form an edge on workbenches to keep bolts, bearings and other small parts from rolling off