Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig SpillBlocker Vacuum Dike
SDP

Catalog No. 17100913 Shop All New Pig Corporation Products
Change view
Click to view available options
Dimensions (L x W x H):
5 x 4 x 2.25 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
17-100-913 5 x 4 x 2.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-913 Supplier New Pig Supplier No. PLR280BK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Attach this unique spill dike to your wet vac system to capture and contain liquids at the same time.

  • Attach this unique spill dike to your wet vac system to capture and contain liquids at the same time.
  • Simply place the dike on the floor and connect it to your wet/dry vacuum
  • Four intakes and integrated vacuum channels suck up liquid quicker than a traditional vacuum head
  • Ideal for spill response or to contain and capture liquid during routine washdowns
  • PVC outlet pipe attaches to most wet vacuum hoses; includes two reducing couplers to fit 1.5 in. and 2.25 in. hoses
  • Connects to any 2.25 in. H SpillBlocker Dike (connector sold separately) to create larger containment area
  • Reusable dikes clean easily with soap and water
  • Polyurethane construction resists water, oils and many chemicals

Specifications

Color Black
Material Polyurethane
Type Spill Blocker
For Use With (Equipment) Wet Vacuum
Packaging Quantity 1 Ea.
Dimensions (L x W x H) 5 x 4 x 2.25 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.