Learn More
Labconco™ SpillStopper™ Work Surfaces for 5 Ft. Protector™ Premier™ and Protector™ XL Hoods
SDPSCI_ED
Catalog No. S28230 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
No Cutouts
Catalog No. Includes
S28230 No Cutouts
1 options
Catalog No. S28230 Supplier Labconco™ Supplier No. 9500500/EMD
Add to Cart
Add to Cart
SpillStopper Work Surfaces are molded of corrosion-resistant solid epoxy. They are contoured to conform to the interior liner of Protector Premier and XL Hoods.
  • Molded from a special formulation of corrosion-resistant epoxy resin
  • Dished surface to contain spills
  • Contoured to conform to the interior liner of Labconco Protector Laboratory Hoods
  • Front edge has a large radius to aerodynamically direct airflow into the hood
  • Pre-drilled 2.5 in. (6.4cm) diameter holes for plumbing pass-through
  • Pre-drilled 1.5 in. (5.8cm) diameter holes for venting
  • May be ordered with pre-cut 6 x 3 in. (15.2 x 7.6cm) oval cupsink cutout(s) in left rear, right rear, left side, or right side. Cupsink is sold separately
Order Info
Service fixture and cup sink sold separately
Specifications
For Use With (Equipment) Protector XL Laboratory Hood
Includes No Cutouts
Product Type Work Surface
Material Epoxy Resin
For Use With (Application) Laboratory Fume Hood Work