Learn More
Corning™ Spin-X™ Centrifuge Tube Insert Without Membrane
Catalog No. 07200120 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
2 mL
Catalog No. Volume (Metric)
07-200-120 2 mL
1 options
This item is not returnable. View return policy
Catalog No. 07-200-120 Supplier Corning™ Supplier No. 9301
Add to Cart
Add to Cart
This item is not returnable. View return policy
Nonsterile and certified nonpyrogenic
  • Spin-X centrifuge tube filters consist of a membrane-containing filter unit within a microcentrifuge tube.
  • Uses: Removing bacteria, cells, and particles from liquids; HPLC sample preparation; DNA removal from agarose or acrylamide gels. Maximum RCF is 16,000 x g.
Order Info
Insert only (microfuge tube not included).
TRUSTED_SUSTAINABILITY
Specifications
Sterility Non-sterile
Volume (Metric) 2 mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov