Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPIN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560749 Supplier Invitrogen™ Supplier No. PA560749
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSPPTVPPMG VDGVSAYLMK KRHTHRKQRR KPTFLTRRNI VG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 43%.

Exhibits H3K4me3-binding activity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPIN4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPIN4
Gene Accession No. Q56A73
Gene Alias 9630042H07Rik; FLJ44984; MGC133224; novel Spin/Ssty repeat containing protein; Spin4; spindlin family member 4; spindlin family, member 4; spindlin-4; TDRD28
Gene Symbols SPIN4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPIN4. Recombinant protein control fragment (Product #RP-96589).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139886
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.