Learn More
Stoelting™ Spinal Adaptor and Clamps
SDP
Catalog No. 10000185 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Spinal Adaptor
Transverse Clamps, Set of 4
Animal:
Mouse
Rat
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Animal
10-000-185 Transverse Clamps, Set of 4 Mouse
10-000-184 Spinal Adaptor Mouse
10-000-186 Spinal Adaptor Rat
10-000-187 Transverse Clamps, Set of 4 Rat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-000-185 Supplier Stoelting™ Supplier No. 51691
Add to Cart
Add to Cart
Holds mice or rats for spinal surgery
  • Design inspired by the need for a versatile instrument for stabilizing the spinal column
  • Accessories to be mounted on the 51600, 51900 or other brands of stereotaxic instruments, converting them for precise spinal surgery applications.
  • Utilizes the manipulator and base plate of the stereotaxic instrument, enabling precision spinal preparations
  • Spinal column is secured by up to four independent transverse process clamps, as well as a spinous process clamp
  • Transverse process clamps are designed to minimize trauma while providing motionless support
  • Engineered for one-handed control of gross and fine adjustment of the surgical preparation in the AP, ML, and DV planes.
  • Transverse process clamps are available to accommodate either the rat or the mouse
Specifications
For Use With (Application) Neuroscience
Animal Mouse
Model Transverse Clamps, Set of 4