Learn More
Spinal Diluting Fluid, Crystal Violet-Acetic Acid Solution for Cerebrospinal Fluid Cell Count, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Physical Form | Solution |
| Chemical Name or Material | SPINAL DILUTING FLUID Crystal Violet- Acetic Acid Solution |
| CAS | 7732-18-5, 64-19-7, 548-62-9 |
| CAS Min % | 90.0, 7.88, 0.1 |
| CAS Max % | 94.0, 8.2, 0.11 |
| Packaging | Natural Poly Bottle |
| Color | Dark Violet |
| pH | <1 |
| Quantity | 500 mL |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.