Description
- 5X enlarged model for detailed examination
- Sectioned both longitudinally and in cross-section to show the anterior and posterior nerve roots, spinal ganglion and blood vessels
- Additional depiction of the spinal cord in cross section through the white and gray matter for more in-depth learning
- Mounted on plastic base
Specifications
Specifications
| Material | Plastic |
| Gender | Male, Female |
| Product Type | Spinal Cord Model |
| Grade Level | High School |