Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spinel Substrate, 10 x 10 x 0.5mm, Polished One Side, 100 Orientation, Thermo Scientific™

Catalog No. AA38505KT
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38505KT 1 Ea.
AA38505KU 5 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA38505KT Supplier Thermo Scientific Chemicals Supplier No. 38505KT

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

100 Orientation

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Spinel Substrate
Name Note Polished One Side
Physical Form Plate
Quantity 1 Ea.
Molecular Formula MgAl2O4
MDL Number MFCD00049475
Synonym Magnesium aluminum oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.