Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPIRE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561960
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561960 Supplier Invitrogen™ Supplier No. PA561960
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HIPVYTLGFE SPQRVSAAKT APIQRRDIFQ SLQGPQWQSV EEAFPHIYSH GCVLKDVCSE CTSFVADVVR SSRKSVDVLN TTPRRSRQTQ SLYIPNTRTL D Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.

Acts as an actin nucleation factor, remains associated with the slow-growing pointed end of the new filament (PubMed:21620703). Involved in intracellular vesicle transport along actin fibers, providing a novel link between actin cytoskeleton dynamics and intracellular transport. Required for asymmetric spindle positioning and asymmetric cell division during meiosis (PubMed:21620703). Required for normal formation of the cleavage furrow and for polar body extrusion during female germ cell meiosis (PubMed:21620703). Also acts in the nucleus: together with SPIRE1 and SPIRE2, promotes assembly of nuclear actin filaments in response to DNA damage in order to facilitate movement of chromatin and repair factors after DNA damage (PubMed:26287480). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPIRE2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPIRE2
Gene Accession No. Q8WWL2
Gene Alias BC026502; KIAA1832; Protein spire homolog 2; SPIR2; spir-2; spire actin nucleation factor 2; spire family actin nucleation factor 2; spire homolog 2; spire homolog 2 (Drosophila); spire type actin nucleation factor 2; SPIRE2; spire-type actin nucleation factor 2
Gene Symbols SPIRE2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPIRE2. Recombinant protein control fragment (Product #RP-98593).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84501
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.