Learn More
Medchemexpress LLC 2,2',7,7'-tetrakis(N,N-di-p-methoxyphenylamino)-9,9'-spirobifluorene | 207739-72-8 | MFCD12022511 | 1225.43 g/mol | C81H68N4O8 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW025445100MG
Encompass_Preferred
Spiro-MeOTAD is a spirobifluorene-based hole-transport layer material used in organic and perovskite solar cells. It facilitates efficient hole extraction and transport in optoelectronic devices and is supplied for research applications; follow supplier guidance for storage and handling.
Catalog No. 50-003-01613