Learn More
Medchemexpress LLC 2,2',7,7'-tetrakis(N,N-di-p-methoxyphenylamino)-9,9'-spirobifluorene | 207739-72-8 | MFCD12022511 | 99.0% | 1225.43 g·mol⁻¹ | C81H68N4O8 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW025445
Spiro-MeOTAD is a high-performance hole-transport layer material used in highly efficient perovskite solar cells.
- High-performance hole-transport layer material.
- Used in highly efficient perovskite solar cells.
- Molecular formula: C81H68N4O8.
- Molecular weight: 1225.43 g·mol⁻¹.
- Available pack sizes include 100 mg, 250 mg, 500 mg, 1 g, 5 g, and 10 g.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.