Learn More
Cayman Chemical SpIronlacton-d3 1mg
Supplier: Cayman Chemical 401241 MG
A TRPA1 antagonist (IC50 0.131 UM in a calcium mobilization assay) selective for human TRPA1 over human TRPV1 and TRPV4 rat TRPV1 TRPV3 and TRPM8 and a panel of 40 additional targets at 10 UM reduces the number of AITC-induced tail flinches in rats at 10 and 30 mg/kg
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.