Learn More
Bachem Splenopentin Acetate
SDP
Supplier:  Bachem 4012856.0025
Encompass_Preferred
Splenopentin Acetate, H-Arg-Lys-Glu-Val-Tyr-OH, 75957-60-7, C31H51N9O9, Mr 693.80, 25mg. SP-5. Splenopentin, RKEVY, is an immunomodulatory pentapeptide corresponding to the amino acid residues 32-36 of the splenic hormone splenin.
Catalog No. 50-260-132
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.