Learn More
Medique Splinter Probe
Catalog No. 19090722 Shop All Medique Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-090-722 1 Ea.
1 options
Catalog No. 19-090-722 Supplier Medique Supplier No. 71601
Add to Cart
Add to Cart
For removing splinters.
  • Stainless-steel construction
  • With sharp triangular blade
  • Autoclavable
Specifications
Product Type Splinter Probe
For Use With (Application) For removing splinters
Length (English) 4 in.
Autoclavable Autoclavable
Design Triangular Blade
Material Stainless Steel
Sterility Sterile
Quantity 1 Ea.