Learn More
Active Motif Splitomicin
SDP
Supplier:  Active Motif 14087
Encompass_Preferred
Active Motif's Splitomicin is a Sirtuin inhibitor that was initially shown to inhibit Sir2p (the yeast Sirt1 homolog) and create a conditional phenocopy of a sir2 deletion mutant in S. cerevisiae. Cells grown in the presence of the drug have silencing defects at telomeres, silent mating-type loci and the ribosomal DNA. Splitomicin inhibits all class III HDACs (sirtuins) and alleviates gene silencing in Fragile X mental retardation syndrome. It also reverses ischemic preconditioning-induced cardioprotection in a mouse model.
Catalog No. NC1840905
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.