Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm
For Gases and Low Molecular Weight Compounds (MW 30-225)
Supplier:  MilliporeSigma™ Supelco™ 57318
 View more versions of this product
Catalog No. 11-100-8581
Only null left
Add to Cart
Description
Description
Solid phase microextraction (SPME) technique is generally used for GC/MS analysis. It helps in simplifying and speeding up the sample preparation. It also reduces solvent requirement during extraction.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SPME
SPME Fibers and Accessories
24 ga
Manual Holder
Fused Silica Fiber
3/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.