Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm
For Gases and Low Molecular Weight Compounds (MW 30-225)
Supplier: MilliporeSigma™ Supelco™ 57335U
Description
Solid phase microextraction (SPME) technique is generally used for GC/MS analysis. It helps in simplifying and speeding up the sample preparation. It also reduces solvent requirement during extraction.
Specifications
| SPME | |
| SPME Fibers and Accessories | |
| 24 ga |
| Autosampler | |
| StableFlex Fiber | |
| 3/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction