Learn More
MilliporeSigma™ Supelco™ SPME Fiber Holder
Use this holder with SPME fiber assemblies that are designed for automated sampling.
Supplier: MilliporeSigma™ Supelco™ 57330U
Description
An adjustable depth guide positions the fiber for sampling and for correct placement in the heated zone of the GC injection port. The fiber can be locked in the exposed position.
Specifications
| Food and Beverages | |
| Manual Sampling | |
| SPME Fibers and Accessories |