Learn More
MilliporeSigma™ Supelco™ SPME Sampling Stand
Holds eight vials while supporting the SPME syringe for consistent fiber immersion depth. Cat. No. 57333-U accommodates 4 mL vials only; Cat. No. 57357-U accommodates 15 mL vials. Order the 15 mL vial puck (Cat. No. 57358-U) as a replacement for the 15 mL unit, or to use 15 mL vials with the 4 mL unit. Not for use with automated/HPLC fiber holders.
Supplier: MilliporeSigma™ Supelco™ 57357U
Description
Specifications
For use with 15 mL vials | |
Food and Beverages | |
SPME Fibers and Accessories |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.