Learn More
MilliporeSigma™ Supelco™ SPME StableFlex™ Fiber Assortment Kit
Catalog No. 111007318 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Autosampler
Needle Gauge:
23 ga
24 ga
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Needle Gauge
11-100-7318 Autosampler 23 ga
11-100-7733 Autosampler 24 ga
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-7318 Supplier MilliporeSigma™ Supelco™ Supplier No. 57284U
Add to Cart
Add to Cart
SPME StableFlex Fiber Assortment Kit contains one fiber of each: 65 μm PDMS/DVB coating, 50/30 μm DVB/Carboxen/PDMS coating, 85 μm Carboxen/PDMS coating and 85 μm Polyacrylate coating
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SPME
For Use With (Equipment) Autosampler
Type SPME Fibers and Accessories
Material StableFlex Fiber
Needle Gauge 23 ga
StableFlex is a trademark of Sigma-Aldrich Co. LLC