Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Spodopan, Serum-free, Protein-free Culture Medium for Insect-cells, w: L-Glutamine, w: 0.35 g/L NaHCO3 (1000 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P04851000
Encompass_Preferred
PAN-Biotech Spodopan, Serum-free, Protein-free medium for Insect-cells, w: L-Glutamine, w: 0.35 g/L NaHCO3. QUANTITY: 1000 ml. MANUFACTURED BY: Pan-Biotech (Germany). DESCRIPTION: PAN-Biotech Spodopan is a serum-free and protein-free medium for optimized growth of insect cells such as Sf9 and Sf21 (Spodoptera frugiperda) in suspension culture. APPLICATION: Spodopan is suitable for the cultivation of insect cells and the production of recombinant proteins and viruses (Baculovirus expression vector system, BEVS). COMPOSITION: Spodopan contains amino acids, vitamins, salts, trace elements, lipids and growth promoting factors in a formulation optimized for insect cells. It contains no protein or any other components of human or animal origin, only traces of animal-derived components (< 0.2 % w/v) and hydrolysates (< 0.7 % w/v).
Catalog No. 50-238-8552
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.