Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPON2/M spondin/Mindin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981073
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1073 Supplier Bioss Supplier No. BS11064R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cell adhesion protein that promotes adhesion and outgrowth of hippocampal embryonic neurons. Binds directly to bacteria and their components and functions as an opsonin for macrophage phagocytosis of bacteria. Essential in the initiation of the innate immune response and represents a unique pattern-recognition molecule in the ECM for microbial pathogens. Binds bacterial lipopolysaccharide (LPS).

Specifications

Antigen SPON2/M spondin/Mindin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SPON2
Gene Accession No. Q9BUD6
Gene Alias 2310045I24Rik; AI504350; Differentially expressed in cancerous and non-cancerous lung cells 1; DIL1; DIL-1; DKFZp686G21139; FLJ16313; FLJ22401; FLJ34460; F-spondin; Mindin; Mspondin; M-spondin; SPON2; spondin 2; spondin 2, extracellular matrix protein; Spondin-2; UNQ435/PRO866
Gene Symbols SPON2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SPON2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10417
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.