Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Sponge-Stick, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. S26783 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Saturation Solution:
10 mL letheen broth
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Saturation Solution
S26783 10 mL letheen broth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S26783 Supplier 3M™ Supplier No. SSL10LET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uses simplified packaging to reduce waste and minimize handling and preparation

  • Stick-mounted sponge design allows access hard to reach areas without directly handling sponge
    • Easily snap off sponge from handle after swabbing
  • 1.5 x 3 in. biocide-free sponge does not affect organism viability
  • 6 x 11 in. sample bag with a 30 oz./0.89 L capacity included for safe, convenient transportation to lab

Specifications

Product Type Sponge-Stick
Length (English) 3 in.
Length (Metric) 7.6 cm
Width (English) 1.5 in.
Width (Metric) 3.8 cm
Packaging Quantity 100/Cs.
Saturation Solution 10 mL letheen broth
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.