Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPOP Recombinant Rabbit Monoclonal Antibody (K01_1Y71)
GREENER_CHOICE

Catalog No. MA563557
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA563557 100 μL
MA565016 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA563557 Supplier Invitrogen™ Supplier No. MA563557
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

SPOP Recombinant Monoclonal Antibody for Western Blot

Component of a cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex that mediates the ubiquitination of target proteins, leading most often to their proteasomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPOP
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Y71
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene SPOP
Gene Accession No. O43791
Gene Alias AI315626; BTBD32; HIB homolog 1; Pcif1; PDX-1 C-terminal-interacting factor 1; roadkill homolog 1; similar to speckle-type POZ protein; speckle type BTB/POZ protein; speckle type POZ protein; speckle-type POZ protein; Spop; SPOP1; TEF2; zgc:64140
Gene Symbols SPOP
Host Species Rabbit
Immunogen A synthetic peptide of human SPOP.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8405
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.