Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Spouting Cylinder
SDPSCI_ED

Catalog No. S43685 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S43685 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43685 Supplier United Scientific Supplies Supplier No. CYSP01
Add to Cart
Add to Cart

Demonstrate Torricelli’s theorem to show the relationship between water pressure and depth.

  • Demonstrate Torricelli's theorem to show the relationship between water pressure and depth.
  • This clear acrylic cylinder, 19-5/16 in. high, includes three vertically aligned spouting nozzles positioned 4-3/4 in. apart.
  • Each spouting nozzle also includes a stopper so pressure at each height can be shown individually.
  • A filler hose barb at the top can be used to quickly refill the cylinder or maintain a constant water depth.

Specifications

Grade 9 to 12
Format Kit
Product Type Spouting Cylinder
Height (English) 19-5/16 in.
Material Acrylic Cylinder
Languages English
Subject Physics - Matter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.