Learn More
Essendant SPRAYANTI-STATIC8OZ
Supplier:  Essendant END096000
Encompass_Preferred
Versatile spray cleans all monitor-screen types as well as just about every surface in your office No-run formula stays where you spray it Alcohol-free to safely clean all of your valuable electronics- tablets televisions computers smartphones and more Removes dust and fingerprints while helping to prevent static-charge build-up Dries streak free For best results use with XL microfiber wipes Application- Electronics/Antistatic Cleaner Applicable Material- Ceramics Fiberglass Glass Laminates Metal Plastic Dirt Types- Debris Dust Grease Fingerprints Soil Smudges Scent- Unscented
Catalog No. 50-000-35385
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.