Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ Spring Clamps for Mighty Small™ Electrophoresis Units
SDP

Catalog No. 03500484
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
SE 250, SE 260 Mini Vertical Electrophoresis Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
03-500-484 SE 250, SE 260 Mini Vertical Electrophoresis Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-500-484 Supplier Hoefer™ Supplier No. SE252
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Mighty Small electrophoresis units

  • Multiple Gel Casters Replacement Parts
  • Some are Asymetrical Clamps

Specifications

Color Red
For Use With (Equipment) SE 250, SE 260 Mini Vertical Electrophoresis Unit
Type Spring Clamp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.