Learn More
Synthware™ Springs
GREENER_CHOICESDP
Catalog No. 31501918 Shop All Kemtech Products
Change view
Click to view available options
Length (Metric):
6.35 mm
25 mm
32 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
31-501-918 6.35 mm
31-501-919 25 mm
31-501-920 32 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-501-918 Supplier Synthware™ Supplier No. TH006
Add to Cart
Add to Cart
Stainless Steel springs for use with glassware that features hooks.
Specifications
Type Spring
Material Stainless Steel
For Use With (Application) Organic Synthesis
Length (Metric) 6.35 mm