Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sprout & Grow™
SCI_ED

Catalog No. S95675 Shop All Educational Insights Products
Change view
Click to view available options
Includes:
Bean and pea seed packets, enriched soil for optimum growth, step-by-step instructions, suggested activities and journal pages for writing about observations
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S95675 Bean and pea seed packets, enriched soil for optimum growth, step-by-step instructions, suggested activities and journal pages for writing about observations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog No. S95675 Supplier Educational Insights Supplier No. EI5101
Add to Cart
Add to Cart
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Students watch seeds sprout and grow into plants — right before their eyes.

Sprout & Grow Greenhouse (S95674)

  • Self-contained kit includes greenhouse with transparent lid, two bags of Wonder Soil (100 mini wafers), three removable six-section trays and activity guide with reproducible worksheet
  • Dimensions (L × W × D): 5.5 × 22.5 × 6.5 in. (14 × 57.2 × 16.5cm)
Sprout & Grow Window (S95675)
  • Allows gardeners of all ages to watch seeds sprout and grow into plants
  • Large, transparent window stands upright on a tabletop or windowsill
  • Special potting soil nurtures seeds for optimum growth
  • Comes with packets of bean and pea seeds and a step-by-step guide with instructions, suggested activities and journal pages
Sprout & Grow Wonder Soil (S95676)
  • Perfect for home and school
  • Wonder Soil mini wafers expand quickly with water and are polymer-fortified for better drainage and faster plant growth

Order Info

Grades PreK+.

Specifications

Color Transparent
For Use With (Application) Perfect for home and school
Includes Bean and pea seed packets, enriched soil for optimum growth, step-by-step instructions, suggested activities and journal pages for writing about observations
Product Type Window™ Planter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.