Learn More
Invitrogen™ SPRR2A Polyclonal Antibody

Description
SPRR2A Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | SPRR2A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | SPRR2A |
| Gene Accession No. | P35326 |
| Gene Alias | 1-Feb; 2-1; small proline rich protein 2A; small proline-rich protein 2A; SPR-2A; SPRR2A |
| Gene Symbols | SPRR2A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.