Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPRY3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. p-200126517
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119434 200 μL
PIPA5121579 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119434 Supplier Invitrogen™ Supplier No. PA5119434
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPRY3 Polyclonal Antibody for Western Blot, IHC (P)

May function as an antagonist of fibroblast growth factor (FGF) pathways and may negatively modulate respiratory organogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRY3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SPRY3
Gene Accession No. O43610
Gene Alias antagonist of FGF signaling; Gm1409; Gm391; Protein sprouty homolog 3; RGD1562172; sprouty homolog 3; sprouty homolog 3 (Drosophila); sprouty RTK signaling antagonist 3; sprouty3; SPRY3; spry-3
Gene Symbols SPRY3
Host Species Rabbit
Immunogen Recombinant protein Sprouty Homolog 3. The antigen corresponds to amino acid range 91-288 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10251, 236576, 498159
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.