Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPRYD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562794 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562794 Supplier Invitrogen™ Supplier No. PA562794
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunogen sequence: FARSLRLCRW GAKRLGVAST EAQRGVSFKL EEKTAHSSLA LFRDDTGVKY GLVGLEPTKV ALNVERFREW AVVLADTAVT SGRHY Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

The SPRY domain-containing protein 4 (SPRYD4) is a member of a family of proteins whose sole common characteristic is the presence of a SPRY domain. SPRY domains are structural domains that were first described in the fungal Dictyostelium discoideum tyrosine kinase spore lysis A. In most systems SPRY domains provide binding sites for regulatory proteins or intramolecular binding sites that maintain the structural integrity of a protein. SPRYD4 is ubiquitously expressed and is most abundant in kidney, brain, bladder, thymus and stomach. Little is known of the function of the SPRYD4 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRYD4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPRYD4
Gene Accession No. Q8WW59
Gene Alias 4633402N23Rik; AA114801; RGD1306649; SPRY domain containing 4; SPRY domain-containing protein 4; Spryd4
Gene Symbols SPRYD4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPRYD4. Recombinant protein control fragment (Product #RP-97853).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283377
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.