Learn More
Seca Corp 535 Spot Check Vital Signs Monitor (NiBP, SpO2 Masimo)
Supplier: Seca Corp 5351200131
The Spot-check-monitor to measure three vital signs: blood pressure, SpO2, and pulse rate. Touchscreen monitor. Large internal memory. EMR-validated: transmit measured data directly to any EMR system. • Measurement of blood pressure, Masimo SpO2, and pulse • 7-inch touchscreen monitor • Internal memory for up to 70,000 measurements • Integrated lithium-ion battery in the monitor • EMR-validated: transmit measured data directly to any EMR system • Connectivity: Wi-Fi, Ethernet • Ports: 2 x USB 2.0 • Software: seca connect 103 • Component options • Temperature probe (oral/axillary) or temperature probe (rectal) • Weight: 5.9 lbs • Dimensions (WxHxD) 9.9 x 10.3 x 10.9 inches • Accessories • Rolling stand seca 475 • Carrying case seca 432 • Sphygmomanometer cuff, sizes X-Small thru XX-Large • Cuff extension tubes seca items 490003301 (51" / 1.5 m) and 490003401 (10 ft / 3 m) • Consumables • Probe covers (oral/axillary, rectal) seca 4900015001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.