Learn More
Seca Corp 535 Spot Check Vital Signs Monitor (NiBP, SpO2 Masimo, Rectal Temp)
SDP
Supplier:  Seca Corp 5351220131
Encompass_Preferred
The Spot-check-monitor to measure four vital signs: blood pressure, SpO2, rectal temp and pulse. Touchscreen monitor. Large internal memory. EMR-validated: transmit measured data to any EMR system. • Measurement of blood pressure, Masimo SpO2, rectal temp, pulse • Easy-to-understand graphical presentation of the measurement results on the touchscreen monitor • Large internal memory for up to 70,000 measurements • EMR-validated to any EMR system• Display: 7-inch touchscreen • Connectivity: Wi-Fi, Ethernet • Ports: 2 x USB 2.0 • Software: seca Connect 103 • Component options • Temperature probe (oral/axillary) • Weight: 5.9 lbs • Dimensions (WxHxD) 9.9 x 10.3 x 10.9 inches • Accessories • Rolling stand seca 475 • Carrying case seca 432
Catalog No. NC1834744