Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPTB Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144546
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144546 Supplier Invitrogen™ Supplier No. PA5144546
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPTB Polyclonal Antibody for Western Blot, ELISA

SPTB (Spectrin Beta, Erythrocytic) is a Protein Coding gene. Diseases associated with SPTB include Spherocytosis, Type 2 and Elliptocytosis 3. Among its related pathways are Cytokine Signaling in Immune system and RET signaling. Gene Ontology (GO) annotations related to this gene include protein heterodimerization activity and actin filament binding. An important paralog of this gene is SPTBN1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPTB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene beta-Spec
Gene Accession No. Q00963
Gene Alias HSPTB1; Spectrin Beta Erythrocytic; Spectrin Beta, Erythrocytic; SPTB1
Gene Symbols beta-Spec
Host Species Rabbit
Immunogen Recombinant Drosophila melanogaster (Fruit fly) beta-Spec protein (1-271aa).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 32746
Target Species Drosophila
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.