Learn More
Epigentek Inc SPTLC2 Polyclonal Antibody (100 ul)
SDP
Supplier:  Epigentek Inc A50500100
Encompass_Preferred
HRP Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Human Serine palmitoyltransferase 2 protein (88-562AA). Tested Applications: ELISA. Reactivity: Human. Alternative Names: Long chain base biosynthesis protein 2, LCB 2, Long chain base biosynthesis protein 2a, LCB2a, Serine-palmitoyl-CoA transferase 2, SPT 2, SPTLC2, KIAA0526, LCB2.
Catalog No. 50-119-6948
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.